Lactulose
Supplier: Apollo Scientific
Warning
BIL9700-100G BIL9700-250G BIL9700-500G
APOSBIL9700-100GEA 277 EUR
APOSBIL9700-100G APOSBIL9700-250G APOSBIL9700-500G
Lactulose
Lactulose
Formula: C₁₂H₂₂O₁₁
MW: 342.297 g/mol
Melting Pt: 169 °C
Storage Temperature: Ambient
MDL Number: MFCD00151469
CAS Number: 4618-18-2