Moniliformin sodium salt Fusarium moniliforme
Supplier: Apollo Scientific
Danger
BIM1004-1MG BIM1004-5MG
APOSBIM1004-1MGEA 574 EUR
APOSBIM1004-1MG APOSBIM1004-5MG
Moniliformin sodium salt Fusarium moniliforme
Moniliformin sodium salt
Formula: C₄HO₃·Na
MW: 120.1 g/mol
Melting Pt: 345…355 °C (dec.)
Storage Temperature: Refrigerator
CAS Number: 71376-34-6
UN: 2811
ADR: 6.1 II