2-Fluoro-2'-deoxyadenosine
Supplier: Apollo Scientific
Synonyms: 2'-Deoxy-2-fluoroadenosine
BIMN2009-100MG BIMN2009-1G BIMN2009-250MG
APOSBIMN2009-100MGEA 132 EUR
APOSBIMN2009-100MG APOSBIMN2009-1G APOSBIMN2009-250MG
2-Fluoro-2'-deoxyadenosine
2-Fluoro-2'-deoxyadenosine
Melting Pt: 218 °C (dec.)
Storage Temperature: Refrigerator
CAS Number: 21679-12-9