1-(4-Nitrophenyl)glycerol
Supplier: Apollo Scientific
Warning
Synonyms: PNPG, 1-(4-Nitrophenyl)-1,2,3-propanetriol
BIN0342-1G BIN0342-2G
APOSBIN0342-1GEA 109 EUR
APOSBIN0342-1G APOSBIN0342-2G
1-(4-Nitrophenyl)glycerol
1-(4-Nitrophenyl)glycerol
Formula: C₉H₁₁NO₅
Melting Pt: 93…101 °C
Storage Temperature: Freezer
MDL Number: MFCD00017037
CAS Number: 2207-68-3
EINECS: 218-624-9