Pyridoxine hydrochloride ≥99.5%
Supplier: Apollo Scientific
Warning
Synonyms: Adermine hydrochloride, Vitamin B6 hydrochloride, PN HCl, Pyridoxine HCl, (5-Hydroxy-6-methylpyridine-3,4-diyl)dimethanol hydrochloride, Pyridoxol hydrochloride, Pyridoxine HCI
BIP0612-100G BIP0612-500G BIP0612-250G
APOSBIP0612-100GEA 116 EUR
APOSBIP0612-100G APOSBIP0612-500G APOSBIP0612-250G
Pyridoxine hydrochloride ≥99.5%
Pyridoxine hydrochloride
Formula: C₈H₁₂ClNO₃
MW: 205.64 g/mol
Melting Pt: 214…215 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00012807
CAS Number: 58-56-0
EINECS: 200-386-2