L-Phenylalanine ≥99.5%
Supplier: Apollo Scientific
Warning
Synonyms: H-L-Phe-OH, Phenylalanine, H-Phe-OH, L-α-Phenylalanine, (S)-(-)-Phenylalanine, 3-Phenyl-L-alanine, 3-Phenyl-L(-)-alanine
BIP0716-100G BIP0716-1KG BIP0716-500G
APOSBIP0716-100GEA 21.9 EUR
APOSBIP0716-100G APOSBIP0716-1KG APOSBIP0716-500G
L-Phenylalanine ≥99.5%
L-α-Phenylalanine
USP, Pyrogen free
Formula: C₉H₁₁NO₂
MW: 165.19 g/mol
Melting Pt: 270…275 °C
Storage Temperature: Ambient
MDL Number: MFCD00064227
CAS Number: 63-91-2
EINECS: 200-568-1