Amino-PEG2-acid tert-butyl ester
Supplier: Apollo Scientific
Warning
Synonyms: tert-Butyl 3-[2-(2-Aminoethoxy)ethoxy]propanoate, 3-[2-(2-Aminoethoxy)ethoxy]propanoic acid tert-butyl ester, 9-Amino-4,7-dioxanonanoic acid tert-butyl ester, NH2-PEG2-C2-Boc, tert-Butyl 9-Amino-4,7-dioxanonanoate, Amino-dPEG®2-t-butyl ester
BIPG1044-250MG BIPG1044-1G
APOSBIPG1044-250MGEA 37.7 EUR
APOSBIPG1044-250MG APOSBIPG1044-1G
Amino-PEG2-acid tert-butyl ester
Amino-PEG2-acid tert-butyl ester
Formula: C₁₁H₂₃NO₄
MW: 233.305 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11041135
CAS Number: 756525-95-8