2-[2-(2-Aminoethoxy)ethoxy]ethanol 97%
Supplier: Apollo Scientific
Danger
BIPG1045-1G BIPG1045-5G BIPG1045-10G
APOSBIPG1045-1GEA 23.2 EUR
APOSBIPG1045-1G APOSBIPG1045-5G APOSBIPG1045-10G
2-[2-(2-Aminoethoxy)ethoxy]ethanol 97%
2-[2-(2-Aminoethoxy)ethoxy]ethanol
Formula: C₆H₁₅NO₃
Density: 1.0773 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD07367495
CAS Number: 6338-55-2
UN: 2735
ADR: 8,II