tert-Butyl 12-Hydroxy-4,7,10-trioxadodecanoate
Supplier: Apollo Scientific
Synonyms: 3-[2-[2-(2-Hydroxyethoxy)ethoxy]ethoxy]propionic acid tert-Butyl ester, Hydroxy-PEG3-(CH2)2-Boc, 12-Hydroxy-4,7,10-trioxadodecanoic acid tert-Butyl ester, tert-Butyl 3-[2-(2-(2-Hydroxyethoxy)ethoxy)ethoxy]propionate
BIPG1449-1G BIPG1449-5G BIPG1449-500MG
APOSBIPG1449-1GEA 102 EUR
APOSBIPG1449-1G APOSBIPG1449-5G APOSBIPG1449-500MG
tert-Butyl 12-Hydroxy-4,7,10-trioxadodecanoate
tert-Butyl 12-Hydroxy-4,7,10-trioxadodecanoate
Formula: C₁₃H₂₆O₆
MW: 278.35 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03791106
CAS Number: 186020-66-6