(Boc-amino)-PEG3-C2-Carboxylic acid
Supplier: Apollo Scientific
Warning
Synonyms: 2,2-Dimethyl-4-oxo-3,8,11,14-tetraoxa-5-azaheptadecan-17-oic acid, Boc-N-amido-PEG3-acid, 12-(Boc-amino)-4,7,10-trioxadodecanoic acid, 12-[(tert-Butoxycarbonyl)amino]-4,7,10-trioxadodecanoic acid
BIPG1801-500MG BIPG1801-100MG BIPG1801-1G
APOSBIPG1801-500MGEA 270 EUR
APOSBIPG1801-500MG APOSBIPG1801-100MG APOSBIPG1801-1G
(Boc-amino)-PEG3-C2-Carboxylic acid
(Boc-amino)-PEG3-C2-Carboxylic acid
Formula: C₁₄H₂₇NO₇
MW: 321.37 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD24539476
CAS Number: 1347750-75-7