Riboflavine (vitamin B2)
Supplier: Apollo Scientific
Warning
Synonyms: Vitamin B2, Lactoflavin, 7,8-Dimethyl-10-((2S,3S,4R)-2,3,4,5-tetrahydroxypentyl)benzo[g]pteridine-2,4(3H,10H)-dione, (-)-Riboflavin, Vitamine G
BIR0613-100G BIR0613-25G
APOSBIR0613-100GEA 29 EUR
APOSBIR0613-100G APOSBIR0613-25G
Riboflavine (vitamin B2)
Riboflavine (Vitamin B2)
Formula: C₁₇H₂₀N₄O₆
MW: 376.37 g/mol
Melting Pt: 290 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00005022
CAS Number: 83-88-5
EINECS: 201-507-1
Merck Index: 12,08367