Bedaquiline
Supplier: Apollo Scientific
Synonyms: (1R,2S)-1-(6-Bromo-2-methoxyquinolin-3-yl)-4-(dimethylamino)-2-(naphthalen-1-yl)-1-phenylbutan-2-ol
BISN0131-5MG BISN0131-10MG
APOSBISN0131-5MGEA 139 EUR
APOSBISN0131-5MG APOSBISN0131-10MG
Bedaquiline
Bedaquiline
Bedaquiline, a diarylquinoline antibiotic that targets ATP synthase, is effective for the treatment of Mycobacterium tuberculosis infections.
Formula: C₃₂H₃₁BrN₂O₂
MW: 555.5 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22628872
CAS Number: 843663-66-1