SB590885
Supplier: Apollo Scientific
Synonyms: 5-(2-(4-(2-Dimethylaminoethoxy)phenyl)-5-(4-pyridinyl)-1H-imidazol-4-yl)-2,3-dihydro-1H-inden-1-one oxime, 5-(2-(4-(2-(Dimethylamino)ethoxy)phenyl)-5-(pyridin-4-yl)-1H-imidazol-4-yl)-2,3-dihydro-1H-inden-1-one oxime
BISN0159-1MG BISN0159-5MG BISN0159-10MG
APOSBISN0159-1MGEA 65.4 EUR
APOSBISN0159-1MG APOSBISN0159-5MG APOSBISN0159-10MG
SB590885
SB590885
MAPK/ERK Signalling Pathway
Formula: C₂₇H₂₇N₅O₂
MW: 453.54 g/mol
Storage Temperature: Freezer
MDL Number: MFCD16038645
CAS Number: 405554-55-4