AZD-8330
Supplier: Apollo Scientific
Synonyms: 2-((2-Fluoro-4-iodophenyl)amino)-N-(2-hydroxyethoxy)-1,5-dimethyl-6-oxo-1,6-dihydropyridine-3-carboxamide, AZD8330
BISN0298-5MG BISN0298-10MG BISN0298-50MG
APOSBISN0298-5MGEA 174 EUR
APOSBISN0298-5MG APOSBISN0298-10MG APOSBISN0298-50MG
AZD-8330
AZD-8330
MAPK/ERK Signalling Pathway
Formula: C₁₆H₁₇FIN₃O₄
MW: 461.2 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18384989
CAS Number: 869357-68-6