N,N,N',N'-Tetramethylethylenediamine (TEMED)
Supplier: Apollo Scientific
|
Danger
|
Synonyms:
1,2-Bis(dimethylamino)ethane, TMEDA, N,N,N',N'-Tetramethylethylenediamine
BIT7140-100ML
BIT7140-250ML
APOSBIT7140-100MLEA
29
EUR
APOSBIT7140-100ML
APOSBIT7140-250ML
N,N,N',N'-Tetramethylethylenediamine (TEMED)
N,N,N',N'-Tetramethylethylenediamine (TEMED)
Formula:
C₆H₁₆N₂ Boiling Pt: 120…122 °C Melting Pt: –55 °C Density: 0.775 g/cm³ (25 °C) Flash Pt: 20 °C Storage Temperature: Ambient |
MDL Number:
MFCD00008335 CAS Number: 110-18-9 UN: 2372 ADR: 3,II |