Ganciclovir
Supplier: Apollo Scientific
Danger
Synonyms: 9-[[2-Hydroxy-1-(hydroxymethyl)ethoxy]methyl]guanine, 2'-Nor-2'-Deoxyguanosine, BW 759, 2-Amino-9-(((1,3-dihydroxypropan-2-yl)oxy)methyl)-1H-purin-6(9H)-one, RS-21592
BITP1699-100MG BITP1699-1G BITP1699-500MG
APOSBITP1699-100MGEA 156 EUR
APOSBITP1699-100MG APOSBITP1699-1G APOSBITP1699-500MG
Ganciclovir
Ganciclovir
  • Protein
Formula: C₉H₁₃N₅O₄
MW: 255.23 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00870588
CAS Number: 82410-32-0