Vanillic acid
Supplier: Apollo Scientific
Warning
Synonyms: 4-Hydroxy-3-methoxybenzoic acid
BIV1001-100G BIV1001-25G
APOSBIV1001-100GEA 36.4 EUR
APOSBIV1001-100G APOSBIV1001-25G
Vanillic acid
Vanillic acid
Formula: C₈H₈O₄
MW: 168.15 g/mol
Melting Pt: 208…214 °C
Storage Temperature: Ambient
MDL Number: MFCD00002551
CAS Number: 121-34-6
EINECS: 204-466-8