Xanthine
Supplier: Apollo Scientific
Warning
Synonyms: 1H-Purine-2,6(3H,7H)-dione, 3,7-Dihydro-1H-purine-2,6-dione, 2,6-Dihydroxypurine
BIX1002-100G BIX1002-250G BIX1002-25G
APOSBIX1002-100GEA 109 EUR
APOSBIX1002-100G APOSBIX1002-250G APOSBIX1002-25G
Xanthine
Xanthine
Formula: C₅H₄N₄O₂
MW: 152.11 g/mol
Melting Pt: 300 °C
Storage Temperature: Ambient
MDL Number: MFCD00078453
CAS Number: 69-89-6
EINECS: 200-718-6
Merck Index: 12,10193