Aluminium 99.999%
Supplier: Apollo Scientific
Warning
Synonyms: Aluminum
IN1003-1KG IN1003-250G
APOSIN1003-1KGEA 0 EUR
APOSIN1003-1KG APOSIN1003-250G
Aluminium 99.999%
Aluminium
Formula: Al
Boiling Pt: 2467 °C
Melting Pt: 660.1 °C
Density: 2.699 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00134029
CAS Number: 7429-90-5
UN: 3178
ADR: 4.1,III