Magnesium chloride, anhydrous 99%
Supplier: Apollo Scientific
Warning
IN1017-1KG IN1017-500G
APOSIN1017-1KGEA 72.5 EUR
APOSIN1017-1KG APOSIN1017-500G
Magnesium chloride, anhydrous 99%
Magnesium chloride
Formula: MgCl₂
MW: 95.21 g/mol
Boiling Pt: 1412
Melting Pt: 714 °C
Density: 2.32 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011106
CAS Number: 7786-30-3
EINECS: 232-094-6
Merck Index: 13,05684