Ammonium dihydrogen phosphate 99.995%
Supplier: Apollo Scientific
Warning
Synonyms: Ammonium phosphate, monobasic, Ammonium Phosphate Monobasic, Ammonium dihydrogenphosphate
IN1063-100G IN1063-500G
APOSIN1063-100GEA 270 EUR
APOSIN1063-100G APOSIN1063-500G
Ammonium dihydrogen phosphate 99.995%
Ammonium dihydrogen phosphate
Formula: NH₄H₂PO₄
MW: 115.03 g/mol
Melting Pt: 190 °C
Density: 1.803 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003396
CAS Number: 7722-76-1
EINECS: 231-764-5
Merck Index: 13,00547