Barium hydroxide octahydrate 99.99%
Supplier: Apollo Scientific
Danger
IN1198-100G IN1198-25G
APOSIN1198-100GEA 161 EUR
APOSIN1198-100G APOSIN1198-25G
Barium hydroxide octahydrate 99.99%
Barium hydroxide octahydrate
Formula: Ba(OH)₂·8H₂O
MW: 315.47 g/mol
Melting Pt: 78 °C
Density: 2.18 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149152
CAS Number: 12230-71-6
UN: 3262
ADR: 8,II