Cesium hydroxide 50% in aqueous solution 99,9%
Supplier: Apollo Scientific
Danger
IN1453-100G IN1453-500G
APOSIN1453-100GEA 270 EUR
APOSIN1453-100G APOSIN1453-500G
Cesium hydroxide 50% in aqueous solution 99,9%
Cesium hydroxide
Formula: CsOH
MW: 149.91 g/mol
Density: 1.72 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010964
CAS Number: 21351-79-1
EINECS: 244-344-1
UN: 2681
ADR: 8,II