Copper(I) bromide 99%
Supplier: Apollo Scientific
Warning
IN1539-1KG IN1539-250G IN1539-10KG
APOSIN1539-1KGEA 848 EUR
APOSIN1539-1KG APOSIN1539-250G APOSIN1539-10KG
Copper(I) bromide 99%
Copper (I) bromide
Formula: BrCu
MW: 143.45 g/mol
Boiling Pt: 1345 °C
Melting Pt: 504 °C
Density: 4.71 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010969
CAS Number: 7787-70-4
EINECS: 232-131-6
Merck Index: 13,02689