Copper(I) iodide 98%
Supplier: Apollo Scientific
Danger
Synonyms: Cuprous iodide
IN1559-100G IN1559-1KG IN1559-500G
APOSIN1559-100GEA 24.8 EUR
APOSIN1559-100G APOSIN1559-1KG APOSIN1559-500G
Copper(I) iodide 98%
Copper (I) iodide
Formula: CuI
MW: 190.45 g/mol
Boiling Pt: 1290 °C
Melting Pt: 605 °C
Density: 5.62 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010978
CAS Number: 7681-65-4
UN: 3077
ADR: 9+EHS III
Merck Index: 13,02692