Copper(II) sulphate pentahydrate 99%
Supplier: Apollo Scientific
Danger
APOSIN1591-1KGEA 93.1 EUR
APOSIN1591-1KG APOSIN1591-100G APOSIN1591-250G
Copper(II) sulphate pentahydrate 99%
Copper(II) sulphate pentahydrate
Formula: CuSO₄·5H₂O
MW: 249,69 g/mol
Melting Pt: 110 °C (dec.)
Density: 2,284 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149681
CAS Number: 7758-99-8
EINECS: 231-847-6
UN: 3288
ADR: 6.1,III