Europium(III) oxide 99.99%
Supplier: Apollo Scientific
Warning
IN1789-25G IN1789-5G
APOSIN1789-25GEA 705 EUR
APOSIN1789-25G APOSIN1789-5G
Europium(III) oxide 99.99%
Europium (III) oxide
Formula: Eu₂O₃
MW: 351.92 g/mol
Boiling Pt: 4118 °C
Melting Pt: 2291 °C
Density: 7.42 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010997
CAS Number: 1308-96-9
EINECS: 215-165-6
Merck Index: 14,03904