Gadolinium(III) carbonate hydrate 99.99%
Supplier: Apollo Scientific
APOSIN1816-25GEA
65.5
EUR
APOSIN1816-25G
Gadolinium(III) carbonate hydrate 99.99%
Gadolinium(III) carbonate hydrate
Formula:
Gd₂(CO₃)₃.xH₂O MW: 494,53 g/mol (anhy) Storage Temperature: Ambient |
MDL Number:
MFCD00150240 CAS Number: 38245-36-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...