Holmium(III) oxide 99,9%
Supplier: Apollo Scientific
|
Warning
|
APOSIN1990-50GEA
277
EUR
APOSIN1990-50G
Holmium(III) oxide 99,9%
Holmium(III) oxide
Formula:
Ho₂O₃ MW: 377,86 g/mol Boiling Pt: 3900 °C Melting Pt: 2360 °C Density: 8,36 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011053 CAS Number: 12055-62-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...