Iron(III) bromide, anhydrous
Supplier: Apollo Scientific
Warning
Synonyms: Ferric bromide, Iron tribromide
IN2056-100G IN2056-25G IN2056-5G
APOSIN2056-100GEA 140 EUR
APOSIN2056-100G APOSIN2056-25G APOSIN2056-5G
Iron(III) bromide, anhydrous
Iron (III) bromide
Formula: Br₃Fe
MW: 295.56 g/mol
Melting Pt: 200 °C
Storage Temperature: Ambient
MDL Number: MFCD00016082
CAS Number: 10031-26-2
EINECS: 233-089-1
Merck Index: 12,04060