Iron(III) chloride hexahydrate ACS
Supplier: Apollo Scientific
|
Danger
|
Synonyms:
Iron trichloride hexahydrate, Ferric chloride hexahydrate
IN2065-1KG
IN2065-5KG
APOSIN2065-1KGEA
221
EUR
APOSIN2065-1KG
APOSIN2065-5KG
Iron(III) chloride hexahydrate ACS
Iron (III) chloride hexahydrate
Formula:
FeCl₃·6H₂O Boiling Pt: 280…285 °C Melting Pt: 37 °C Density: 1.82 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00149712 CAS Number: 10025-77-1 UN: 3260 ADR: 8,III |