Ferric pyrophosphate, Technical Grade
Supplier: Apollo Scientific
Warning
Synonyms: Iron(III) pyrophosphate, Iron (III) pyrophosphate
IN2087-1KG IN2087-500G
APOSIN2087-1KGEA 203 EUR
APOSIN2087-1KG APOSIN2087-500G
Ferric pyrophosphate, Technical Grade
Ferric pyrophosphate
Formula: Fe₄O₂₁P₆
MW: 745.2100 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00016091
CAS Number: 10058-44-3