Lithium carbonate 99.99%

Supplier: Apollo Scientific
Danger

APOSIN2317-250GEA 818 EUR
APOSIN2317-250G
Lithium carbonate 99.99%
Lithium carbonate
Formula: Li₂CO₃
MW: 73,89 g/mol
Boiling Pt: 1310 °C
Melting Pt: 720 °C
Density: 2,11 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011084
CAS Number: 554-13-2
EINECS: 209-062-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR