Lithium chloride, anhydrous 99,9%
Supplier: Apollo Scientific
Warning
IN2323-1KG IN2323-250G
APOSIN2323-1KGEA 841 EUR
APOSIN2323-1KG APOSIN2323-250G
Lithium chloride, anhydrous 99,9%
Lithium chloride
Formula: LiCl
MW: 42.39 g/mol
Boiling Pt: 1325…1360 °C
Melting Pt: 605 °C
Density: 2.07 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011078
CAS Number: 7447-41-8
EINECS: 231-212-3