Lithium chloride, anhydrous 99,95%
Supplier: Apollo Scientific
Warning
APOSIN2326-250GEA 448 EUR
APOSIN2326-250G APOSIN2326-25G APOSIN2326-100G
Lithium chloride, anhydrous 99,95%
Lithium chloride
Formula: LiCl
MW: 42,39 g/mol
Boiling Pt: 1325…1360 °C
Melting Pt: 605 °C
Density: 2,068 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011078
CAS Number: 7447-41-8