Magnesium sulphate 98%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
MgSO4, Magnesium sulphate anhydrous
IN2448-1KG
IN2448-500G
APOSIN2448-1KGEA
24.8
EUR
APOSIN2448-1KG
APOSIN2448-500G
Magnesium sulphate 98%
Magnesium sulphate
Formula:
MgSO₄ MW: 120.37 g/mol Melting Pt: 1124 °C (dec.) Density: 2.66 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011110 CAS Number: 7487-88-9 EINECS: 231-296-2 |