Neodymium(III) oxide 99.999%
Supplier: Apollo Scientific
Synonyms: Dineodymium trioxide, Neodymium(III)-oxide, Neodymium oxide
IN2656-100G IN2656-10G
APOSIN2656-100GEA 659 EUR
APOSIN2656-100G APOSIN2656-10G
Neodymium(III) oxide 99.999%
Neodymium (III) oxide
Formula: Nd₂O₃
MW: 336.48 g/mol
Boiling Pt: 3760 °C
Melting Pt: 2272 °C
Density: 7.24 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011134
CAS Number: 1313-97-9
Merck Index: 14,06450