Nickel(II) chloride hexahydrate 99,95%
Supplier: Apollo Scientific
Danger
Synonyms: Nickel dichloride hexahydrate
IN2683-1KG IN2683-250G
APOSIN2683-1KGEA 342 EUR
APOSIN2683-1KG APOSIN2683-250G
Nickel(II) chloride hexahydrate 99,95%
Nickel (II) chloride hexahydrate
Formula: NiCl₂·6H₂O
MW: 237.69 g/mol
Melting Pt: 140
Storage Temperature: Ambient
MDL Number: MFCD00149809
CAS Number: 7791-20-0
UN: 3288
ADR: 6.1 III
Merck Index: 13,06529