Nickel (II) oxide 99%
Supplier: Apollo Scientific
Danger
Synonyms: Nickel monoxide, Nickel oxide, Bunsenite
IN2701-100G IN2701-500G
APOSIN2701-100GEA 132 EUR
APOSIN2701-100G APOSIN2701-500G
Nickel (II) oxide 99%
Nickel (II) oxide
Formula: NiO
MW: 74.69 g/mol
Melting Pt: 1984 °C
Density: 6.67 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011145
CAS Number: 1313-99-1