Nickel (II) oxide 99.99+%
Supplier: Apollo Scientific
Danger
Synonyms: Nickel monoxide, Nickel oxide, Bunsenite
IN2704-100G IN2704-50G
APOSIN2704-100GEA 284 EUR
APOSIN2704-100G APOSIN2704-50G
Nickel (II) oxide 99.99+%
Nickel (II) oxide
Formula: NiO
Melting Pt: 1900 °C
Density: 6.67 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011145
CAS Number: 1313-99-1