Phosphorus(V) bromide 95%
Supplier: Apollo Scientific
Danger
Synonyms: Phosphorus pentabromide
IN2860-100G IN2860-25G
APOSIN2860-100GEA 0 EUR
APOSIN2860-100G APOSIN2860-25G
Phosphorus(V) bromide 95%
Phosphorus (V) bromide
Formula: PBr₅
MW: 430.49 g/mol
Melting Pt: 105…107 °C
Density: 3.61
Storage Temperature: Ambient
MDL Number: MFCD00011437
CAS Number: 7789-69-7
EINECS: 232-186-6
UN: 2691
ADR: 8 II
Merck Index: 12,07507