Potassium nitrate 99,9%
Supplier: Apollo Scientific
Warning
Synonyms: Nitrate of Potash
IN2926-100G IN2926-500G
APOSIN2926-100GEA 0 EUR
APOSIN2926-100G APOSIN2926-500G
Potassium nitrate 99,9%
Potassium nitrate
Formula: KNO₃
Boiling Pt: 400 °C
Melting Pt: 334 °C
Density: 2.109 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011409
CAS Number: 7757-79-1
EINECS: 231-818-8
UN: 1486
ADR: 5.1,III