Discandium trioxide 99.99%
Supplier: Apollo Scientific
Warning
Synonyms: Scandium oxide, Scandium (III) oxide
IN3148-1G IN3148-5G IN3148-25G
APOSIN3148-1GEA 116 EUR
APOSIN3148-1G APOSIN3148-5G APOSIN3148-25G
Discandium trioxide 99.99%
Discandium trioxide
Formula: O₃Sc₂
MW: 137.91 g/mol
Melting Pt: 2485 °C
Density: 3.864 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011223
CAS Number: 12060-08-1