Silicon 99,9%, powder –100 mesh
Supplier: Apollo Scientific
Warning
IN3181-100G IN3181-500G
APOSIN3181-100GEA 87 EUR
APOSIN3181-100G APOSIN3181-500G
Silicon 99,9%, powder –100 mesh
Silicon
Formula: Si
MW: 28.09 g/mol
Boiling Pt: 2600 °C
Melting Pt: 1420 °C
Density: 2.329 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00085311
CAS Number: 7440-21-3
EINECS: 231-130-8
UN: 1346
ADR: 4.1 III
Merck Index: 12,08635