Sodium metabisulphite
Supplier: Apollo Scientific
Warning
APOSIN3247-500GEA 94.5 EUR
APOSIN3247-500G
Sodium metabisulphite
Sodium metabisulphite
Formula: Na₂S₂O₅
MW: 190,11 g/mol
Melting Pt: 150 °C
Density: 1,48 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00167602
CAS Number: 7681-57-4
EINECS: 231-548-0