Sodium tungstate dihydrate 98-101% ACS
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
Tungstic acid sodium salt dihydrate, Sodium wolframate dihydrate, Sodium tungsten oxide dihydrate
IN3275-100G
APOSIN3275-100GEA
72.5
EUR
APOSIN3275-100G
Sodium tungstate dihydrate 98-101% ACS
Sodium wolframate dihydrate
Formula:
Na₂WO₄·2H₂O MW: 329.85 g/mol Melting Pt: 698 °C Density: 3.23…3.25 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00149190 CAS Number: 10213-10-2 EINECS: 236-743-4 |