Strontium bromide, anhydrous 99,9%
Supplier: Apollo Scientific
IN3340-250G
IN3340-50G
APOSIN3340-250GEA
502
EUR
APOSIN3340-250G
APOSIN3340-50G
Strontium bromide, anhydrous 99,9%
Strontium bromide
Formula:
Br₂Sr Melting Pt: 643 °C Density: 4.216 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00049548 CAS Number: 10476-81-0 |