Strontium chloride 99%
Supplier: Apollo Scientific
Danger
IN3352-500G
APOSIN3352-500GEA 66.7 EUR
APOSIN3352-500G
Strontium chloride 99%
Strontium chloride
Formula: Cl₂Sr
MW: 158.53 g/mol
Boiling Pt: 1250 °C
Melting Pt: 873 °C
Density: 3.052 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011249
CAS Number: 10476-85-4
EINECS: 233-971-6