Strontium chloride 99.999%
Supplier: Apollo Scientific
Warning
IN3355-25G IN3355-5G
APOSIN3355-25GEA 261 EUR
APOSIN3355-25G APOSIN3355-5G
Strontium chloride 99.999%
Strontium chloride
Formula: Cl₂Sr
MW: 158.53 g/mol
Melting Pt: 873 °C
Density: 3.05 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011249
CAS Number: 10476-85-4