Sulphur, powder sublimed
Supplier: Apollo Scientific
Warning
IN3391-2KG IN3391-500G
APOSIN3391-2KGEA 0 EUR
APOSIN3391-2KG APOSIN3391-500G
Sulphur, powder sublimed
Sulphur
Formula: S
MW: 32.07 g/mol
Boiling Pt: 445 °C
Melting Pt: 112 °C
Density: 2.07 g/cm³ (25 °C)
Flash Pt: 168 °C
Storage Temperature: Ambient
MDL Number: MFCD00085316
CAS Number: 7704-34-9
UN: 1350
ADR: 4.1 III
Merck Index: 13,09059